Skip to Content
Merck
All Photos(1)

Documents

BI0158

Sigma-Aldrich

Activator Reagent 0.5M ETT in Acetonitrile

0.5M ETT in Acetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116006

Quality Level

200
300

product line

Novabiochem®

form

liquid

reaction suitability

reaction type: Coupling Reactions
reagent type: catalyst

transition temp

flash point 5.6 °C

shipped in

ambient

storage temp.

2-30°C

Application

Ethyl thiotetrazole (ETT) in Acetonitrile can be used as an efficient activator reagent for the synthesis of oligonucleotides. This reagent is effective in the solid-phase synthesis of RNA oligonucleotides in terms of product yield in comparison with other reagents such as 4,5-dicyanoimidazole and 5-benzylthio-1H-tetrazole (BTT).

Analysis Note

Color (APHA): 10 max
ETT [5-(Ethylthio)-1H-tetrazole] Molarity: 0.48-0.52M
Form: Clear liquid
Water (H2O): 60ppm max

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

42.1 °F

Flash Point(C)

5.6 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefanie Hoffmann et al.
Current protocols in nucleic acid chemistry, Chapter 4, Unit 4-Unit 4 (2011-09-09)
This unit describes the solid-phase synthesis and downstream processing for RNA oligonucleotides with a length of up to 40 to 50 nucleotides on a 1- to 4-mmol scale with subsequent conjugation to PEG using the L-RNA spiegelmer NOX-E36 as an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service