Skip to Content
Merck
All Photos(1)

Documents

BI0152

Sigma-Aldrich

Activator Solution 0.25M 5-(Ethylthio)-1H-Tetrazole in Acetonitrile

Biosynthesis Novabiochem®

Synonym(s):

1H-tetrazole

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116006

vapor pressure

97 hPa ( 20 °C)

Quality Level

product line

Novabiochem®

form

liquid

reaction suitability

reaction type: Coupling Reactions
reagent type: catalyst

bp

81.6 °C/1013 hPa (Acetonitrile)

transition temp

flash point 7.2 °C

shipped in

ambient

storage temp.

2-30°C

Application

BI0152-0201 si packaged in 24/400 glass bottles
BI0152-0450 is packaed in 28/405 glass bottles
BI0152-0950 is packaged in 38/430 glass bottles
BI0152-1005 is packaged in GL45 glass bottles
BI0152-2500 is packaged in 38/430 glass bottles
BI0152-2505 is packaged in GL45 glass bottles
BI0152-P4000 is packaged in poly coated glass bottles
DNA Reagents Label Change Explanation
Biosynthesis Novabiochem®

Analysis Note

Color (APHA): 10 max
Component: Acetonitrile Purity (By GC): 99.9% min
ETT [5-(Ethylthio)-1H-tetrazole] Molarity: 0.23-0.27M
Form: Clear liquid
Water (H2O): 30ppm max

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

45.0 °F - closed cup

Flash Point(C)

7.2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service