Skip to Content
Merck
All Photos(1)

Key Documents

AX0149

Supelco

Acetonitrile

≥99.5% (GC), rACS reagent, for extraction, for analytical purposes

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
597,00 kr.
4 L
3.410,00 kr.

597,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
597,00 kr.
4 L
3.410,00 kr.

About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
MDL number:
UNSPSC Code:
12352117
EC Index Number:
200-835-2
grade:
ACS reagent
for extraction
reagent grade
application(s):
environmental
food and beverages
industrial qc
pharmaceutical
sample preparation
bp:
81-82 °C (lit.)
vapor pressure:
72.8 mmHg ( 20 °C)
97 hPa ( 20 °C)

597,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Acetonitrile, for extraction, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

grade

ACS reagent
for extraction
reagent grade

Quality Level

Agency

meets USP/NF

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)
97 hPa ( 20 °C)

description

for analytical purposes

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

973 °F

expl. lim.

16 %

dilution

(for analytical testing)

impurities

≤0.3% Water (H2O)
≤0.6 μeq/g Titrable base
≤8 μeq/g Titrable acid

evapn. residue

≤0.005%

color

clearAPHA: ≤10

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

transition temp

flash point 2 °C

density

0.786 g/mL at 25 °C (lit.)

application(s)

environmental
food and beverages
industrial qc
pharmaceutical
sample preparation

format

neat

shipped in

ambient

storage temp.

room temp

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Analysis Note

AppearanceClearAssay (GC)99.5% minColor (APHA)10 maxEvaporation residue0.005% maxTitrable acid8µeq/g maxTitrable base0.6µeq/g maxWater (H2O)0.3% max

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service