Skip to Content
Merck
All Photos(1)

Documents

AQ112F

Sigma-Aldrich

Goat Anti-Human IgG Antibody, F(ab′)2, FITC conjugate

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

FITC conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

Specificity

Based on immunoelectrophoresis, the antibody reacts with the heavy chains on human IgG and with light chains common to most human immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins, but the antibody may cross-react with immunoglobulins from other species.

Immunogen

purified human IgG

Application

Immunohistochemistry: 1:500-1:5000

EIA and Western blots: 1:5000-1:100,000

Optimal working dilutions must be determined by end user.
Research Category
Secondary & Control Antibodies
Research Sub Category
Fragment Specific Secondary Antibodies
This Goat anti-Human IgG Antibody, F(ab′)2, FITC conjugate is validated for use in IF for the detection of Human IgG.

Linkage

Replaces: AQ316F

Physical form

Affinity purified antibody. Lyophilized. Buffer = 0.01 M Sodium Phosphate, 0.25 M NaCl, pH 7.6 with 15mg/mL BSA, 0.01% thimerosal and 0.05% Sodium Azide

Storage and Stability

Reconstitution: Store freeze-dried product at 4°C until opened for up to 12 months. After opening, restore to 0.8 mL with distilled water to yield 1.4 mg/mL antibody concentration. Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. (To judge clarity, draw product into a pasteur pipette). Product is stable for several weeks at 4°C as an undiluted liquid. Discard dilutions after more than one day. For extended storage after reconsitutiton, add an equal volume of glycerol to make a final concentation of 50% glycerol and store at -20°C. Please remember that the concentration of protein (and buffer salts) will decrease to one-half of the original after addition of glycerol.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yinglan Guo et al.
Science advances, 9(47), eadj2641-eadj2641 (2023-11-24)
Staphylococcus epidermidis expresses glycerol phosphate wall teichoic acid (WTA), but some health care-associated methicillin-resistant S. epidermidis (HA-MRSE) clones produce a second, ribitol phosphate (RboP) WTA, resembling that of the aggressive pathogen Staphylococcus aureus. RboP-WTA promotes HA-MRSE persistence and virulence in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service