Skip to Content
Merck
All Photos(1)

Key Documents

AP128

Sigma-Aldrich

Goat Anti-Mouse IgM Antibody

Goat Anti-Mouse IgM Antibody

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
2.010,00 kr.

2.010,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
2 MG
2.010,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

2.010,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

Application

Goat anti-Mouse IgM Antibody is an antibody against Mouse IgM for use in ELISA, IP & WB.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph F Bonner et al.
Methods in molecular biology (Clifton, N.J.), 1078, 65-88 (2013-08-27)
Neural stem cells (NSC) are not only a valuable tool for the study of neural development and function, but an integral component in the development of transplantation strategies for neural disease. NSC can be used to study how neurons acquire

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service