Skip to Content
Merck
All Photos(1)

Key Documents

ABT268

Sigma-Aldrich

Anti-Liprin Alpha 1

from rabbit, purified by affinity chromatography

Synonym(s):

LAR-interacting protein 1, LIP-1, Protein tyrosine phosphatase receptor type f polypeptide-interacting protein alpha-1, PTPRF-interacting protein alpha-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
2.040,00 kr.

2.040,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
2.040,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

2.040,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... PPFIA1(8500)

General description

Liprin-alpha-1 (UniProt: Q13136; also known as LAR-interacting protein 1, LIP-1, Protein tyrosine phosphatase receptor type f polypeptide-interacting protein alpha-1, PTPRF-interacting protein alpha-1) is encoded by the PPFIA1 (also known as LIP1) gene (Gene ID: 8500) in human. Liprin-alpha-1 is a member of the LAR (leukocyte common antigen-related) protein tyrosine phosphatase-interacting protein (liprin) family. It is a cytoplasmic protein that regulates the disassembly of focal adhesions. It forms homodimers and heterodimers with Liprin-alpha and Liprin-beta. Liprins contain an N-terminal coiled-coil domain and a C-terminal liprin homology (LH) region comprised of three sterile alpha motif (SAM) domains. The N-terminal region acts as a binding region for several synaptic proteins. SAM domain are known to bind to both protein kinases and phosphatases. In mast cells, liprin-alpha-1 facilitates exocytosis and cell spreading.

Specificity

This polyclonal antibody detects Liprin Alpha 1 in human. It targets an epitope within 179 amino acids from the internal region of isoform 1 and 2.

Immunogen

GST-tagged recombinant fragment of 179 amino acids from the internal region of human Liprin Alpha 1.

Application

Research Category
Cell Structure
This rabbit polyclonal Anti-Liprin Alpha 1, Cat. No. ABT268 detects Liprin alpha 1 in human cells. It has been tested for use in Western Blotting.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Liprin Alpha 1 in 10 µg of HeLa cell lysate.

Target description

~160 kDa observed; 135.78 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service