Skip to Content
Merck
All Photos(2)

Key Documents

ABT1890

Sigma-Aldrich

Anti-MTNR1B

from rabbit, purified by affinity chromatography

Synonym(s):

Melatonin receptor type 1B, Mel-1B-R

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
952,00 kr.
100 μG
2.410,00 kr.

952,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
952,00 kr.
100 μG
2.410,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

952,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... MTNR1B(4544)

General description

Melatonin receptor type 1B (UniProt: P49286; also known as Mel-1B-R, Mel1b receptor) is encoded by the MTNR1B gene (Gene ID: 4544) in human. Mel-1B-R is a member of the G-protein coupled 1 family that is predominantly expressed in retina and to lesser extent in brain tissue. . Mel-1B-R activity is mediated by pertussis toxin sensitive G proteins that inhibit adenylate cyclase activity. Mel-1B-R mediates the reproductive and circadian actions of melatonin. Melatonin is shown to be involved in body mass regulation in mammals and this effect is suggest to be mediated by a direct action of melatonin on Mel-1B receptors on brown adipocytes. Melatonin is reported to act as a chemopreventive agent and its levels inversely correlate with the risk of developing cancer. In this regard melatonin induces p38-dependent phosphorylation of both p53 and histone H2AX. Activation of the p53-dependent DNA damage response by melatonin is mediated by both Mel-1A and Mel-1B receptors. The absence of either receptor impairs melatonin′s ability to reduce both cell proliferation and clonogenic potential of cancer cells. A variant in MTNR1B gene is reported to worsen the deleterious effect of melatonin on glucose tolerance in humans. (Ref.: Brydon, L. et al. (2001). Endocrinology. 142(10):4264-4271; Santoro, R., et al. (2013). Carcinogenesis. 34(5):1051-1061).

Specificity

This rabbit polyclonal antibody detects Melatonin receptor type 1B in human.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to 20 amino acids from the N-terminal region of human Melatonin receptor type 1B.

Application

Anti-MTNR1B Antibody, Cat. No. ABT1890, is a rabbit polyclonal antibody that detects MTNR1B and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected MTNR1B in human retina tissue.
Research Category
Cell Structure

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected MTNR1B in 10 µg of HepG2 cell lysate.

Target description

40.19 kDa calculated.

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanliang Li et al.
International journal of molecular medicine, 41(2), 955-961 (2017-12-06)
The pineal hormone melatonin influences the secretion of insulin by pancreatic islets via the G‑protein‑coupled melatonin receptors 1 and 2 that are expressed in pancreatic β‑cells. Genome‑wide association studies indicate that melatonin receptor 1B (MTNR1B) single nucleotide polymorphisms are tightly associated with type 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service