Skip to Content
Merck
All Photos(1)

Key Documents

ABS2248

Sigma-Aldrich

Anti-OVCH2

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
1.210,00 kr.
100 μL
3.070,00 kr.

1.210,00 kr.


Estimated to ship on28 March 2025Details


Request a Bulk Order

Select a Size

Change View
25 μL
1.210,00 kr.
100 μL
3.070,00 kr.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

1.210,00 kr.


Estimated to ship on28 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

mol wt

calculated mol wt 67.61 kDa
observed mol wt ~74 kDa

species reactivity

mouse

packaging

antibody small pack of 100 μL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

dry ice

storage temp.

-10 to -25°C

target post-translational modification

unmodified

Gene Information

mouse ... Ovch2(244199)

General description

Ovochymase-2 (UniProt: Q7M761; Oviductin, OVCH2) is encoded by the Ovch2 (also known as Ovtn) gene (Gene ID: 244199) in murine species. OVCH2 is a member of the peptidase S1 family that is synthesized with a signal peptide (aa 1-22) that is subsequently cleaved off. It also has a propeptide (aa 23-51) that serves as an activation peptide. OVCH2 is mainly localized in the caput region of epididymis that is most proximal to the testis. It is shown to be essential for processing ADAM3, the sperm surface protein that is required for the fertilizing ability of sperm. It is reported that a neural epidermal growth factor like like 2 (NELL2), binds to an orphan receptor tyrosine kinase, c-Ros oncogene 1 (ROS1), and mediates the differentiation of the initial segment (IS) of the caput epididymis. Male mice with NELL2 are reported to be infertile. (Ref.: Kiyozumi, D., et al. (2020). Science. 368(6495); 1132-1135).

Specificity

This rabbit polyclonal antibody detects murine Ovochymase-2 (OVCH2). It targets an epitope within the CUB2 domain from the C-terminal region.

Immunogen

GST-tagged recombinant fragment corresponding to 179 amino acids from the C-terminal region of murine Ovochymase-2 (OVCH2).

Application

Quality Control Testing

Evaluated by Western Blotting in mature mouse caput epididymis lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected OVCH2 in mature mouse caput epididymis lysate.

Tested Applications

Western Blotting Analysis: A representative lot detected OVCH2 in mature mouse caput epididymis lysate (Courtesy of Dr. Masahito Ikawa Lab @ Osaka University, Japan).

Western Blotting Analysis: A representative lot detected OVCH2 in Western Blotting applications (Kiyozumi, D., et. al. (2020). Science. 368(6495):1132-1135).

Immunohistochemistry Applications: A representative lot detected OVCH2 in Immunohistochemistry applications (Kiyozumi, D., et. al. (2020). Science. 368(6495):1132-1135).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
This rabbit polyclonal Anti-OVCH2, Cat. No. ABS2248, is a rabbit polyclonal antibody that detects Ovochymase-2 (OVCH2) and is tested for use in Immunohistochemistry and Western Blotting.

Physical form

Rabbit polyclonal antibody in 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service