Skip to Content
Merck
All Photos(3)

Key Documents

ABS1484

Sigma-Aldrich

Anti-RASSF1

from rabbit, purified by affinity chromatography

Synonym(s):

Ras association domain-containing protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
952,00 kr.
100 μL
2.830,00 kr.

952,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
952,00 kr.
100 μL
2.830,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

952,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat

packaging

antibody small pack of 25 μL

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... RASSF1(11186)

Related Categories

General description

Ras association domain-containing protein 1 (UniProt: Q9NS23; RASSF1) is encoded by the RASSF1 (also known as RDA32) gene (Gene ID: 11186) in human. RASSF1 serves as a potential tumor suppressor and is shown to be essential for death receptor-dependent apoptosis. It mediates the activation of STK3/MST2 and STK4/MST1 during Fas-induced apoptosis by preventing their dephosphorylation. In association with MOAP1, it promotes Bax conformational change and translocation to mitochondrial membranes in response to TNF and TNFSF10 stimulation. Eight isoforms of RASSF1 have been described that are produced by alternative promoter usage and alternative splicing. The immunogen sequence used to generate this antibody is conserved in isoforms A, D, E, F and G. Isoform A is ubiquitously expressed and is shown to localize to cytoplasmic microtubules during interphase, to bipolar centrosomes associated with microtubules during prophase, to spindle fibers and spindle poles at metaphase and anaphase, to the midzone during early telophase, and to the midbody in late telophase in cells. Isoform A is reported to interact with CDC20, an activator of the anaphase-promoting complex (APC), resulting in the inhibition of APC activity and mitotic progression. It Inhibits proliferation by negatively regulating cell cycle progression at the level of G1/S-phase transition by regulating accumulation of cyclin D1 protein. It also disrupts interactions among MDM2, DAXX and USP7 and thereby contributes to the efficient activation of TP53 by promoting MDM2 self-ubiquitination in cell-cycle checkpoint control in response to DNA damage.

Specificity

This rabbit polyclonal antibody detects Ras association domain-containing protein 1 in human and rat tissues. It targets an epitope within 14 amino acids from the N-terminal region.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to 14 amino acids from the N-terminal region of human Ras association domain-containing protein 1.

Application

Anti-RASSF1, Cat. No. ABS1484, is a rabbit polyclonal antibody tht detects Ras association domain-containing protein 1 in human and rat tissues and has been tested for use in Flow Cytometry, Immunohistochemistry (paraffin), and Western Blotting.
Flow Cytometry Analysis: 1 µg from a representative lot detected RASSF1 in Jurkat cells.

Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected RASSF1 in human ovarian cancer, human placenta, and human pancreas tissue sections.
Research Category
Signaling

Quality

Evaluated by Western Blotting in rat liver tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected RASSF1 in 10 µg of rat liver tissue lysate.

Target description

~39 kDa observed; 39.22 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service