Skip to Content
Merck
All Photos(3)

Key Documents

ABN1721M

Sigma-Aldrich

Anti-TMEM100

from rabbit, purified by affinity chromatography

Synonym(s):

Transmembrane protein 100

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
1.120,00 kr.

1.120,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
1.120,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

1.120,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Tmem100(67888)

General description

Transmembrane protein 100 (UniProt: Q9CQG9; also known as TMEM100) is encoded by the Tmem100 gene (Gene ID: 67888) in murine species. TMEM100 is a multi-pass membrane protein that is shown to be involved in renal development, vasculogenesis and lung cancer invasiveness. It plays a role during embryonic arterial endothelium differentiation and vascular morphogenesis through the ACVRL1 receptor-dependent signaling pathway upon stimulation by bone morphogenetic proteins, such as GDF2/BMP9 and BMP10. TMEM100 is also involved in the regulation of nociception by forming a complex with TRPA1 and TRPV1 and act as a modulator of their interaction in dorsal root ganglia (DRG) neurons. It is reported to weaken TRPA1 and TRPV1 interaction, thereby releasing the inhibition of TRPA1 by TRPV1 and increasing the single-channel open probability of the TRPA1-TRPV1 complex. TMEM100-deficient mice show a reduction in inflammatory mechanical hyperalgesia and TRPA1- but not TRPV1-mediated pain. A mutation involving aa 117-119 KRR to QQQ is shown to abolish TMEM100 interaction with TRPA1, but increases interaction with TRPV1. Mutations is Tmem100 gene can lead to embryonic lethality due to impaired differentiation of arterial endothelium and defects of vascular morphogenesis. (Ref.: Weng, H-J., et al. (2015). Neuron 85(4), 833-846).

Specificity

This rabbit polyclonal antibody detects transmembrane protein 100 (TMEM100) in murine cells. It targets an epitope with the 19 amino acids from the N-terminal end.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the N-terminus region of murine Transmembrane protein 100 (TMEM100).

Application

Anti-TMEM100, Cat. No. ABN1721, is a highly specific rabbit polyclonal antibody that targets Transmembrane protein 100 and has been tested in Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.
Immunohistochemistry Analysis: A 1:50-250 dilution from a representative lot detected TMEM100 in mouse colon, rat peripheral nerve, and human cerebral cortex tissues.

Immunofluorescence Analysis: A representative lot detected TMEM100 in DRGs (Weng, H.J., et. al. (2015). Neuron. 85(4):833-46).

Western Blotting Analysis: A representative lot detected TMEM100 in DRG from cervical to lumbar levels (Weng, H.J., et. al. (2015). Neuron. 85(4):833-46).

Immunoprecipitation Analysis: representative lot detected TMEM100 in whole-cell DRG or CHO cells lysates (Weng, H.J., et. al. (2015). Neuron. 85(4):833-46).

Western Blotting Analysis: A 1:5,000 dilution from a representative lot detected TMEM100 in Dorsal root ganglion (DRG) of WT & TMEM100 CKO mice (Courtesy of Drs. Hao-Jui Weng and Xinzhong Dong at Johns Hopkins University).

Immunofluorescence Analysis: A 1:200 dilution from a representative lot detected TMEM100 in Dorsal root ganglion (DRG) sections from Avil-Cre;Tmem100+/+ (Wild Type mice) and Avil-Cre;Tmem100fl/fl (Tmem100 CKO) mice (Courtesy of Drs. Hao-Jui Weng and Xinzhong Dong at Johns Hopkins University).


Quality

Evaluated by Western Blotting in mouse lung tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected TMEM100 in 10 µg of mouse lung tissue lysate.

Target description

~17 kDa observed; 14.50 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service