Skip to Content
Merck
All Photos(4)

Key Documents

ABN1662

Sigma-Aldrich

Anti-STRA6

from rabbit

Synonym(s):

Stimulated by retinoic acid gene 6 protein, Retinoic acid-responsive protein

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
1.120,00 kr.
100 μL
2.790,00 kr.

1.120,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
1.120,00 kr.
100 μL
2.790,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

1.120,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

unpurified

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

packaging

antibody small pack of 25 μL

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Stra6(20897)

General description

Stimulated by retinoic acid gene 6 protein (UniProt: O70491; also known as Retinoic acid-responsive protein, STRA6) is encoded by the Stra6 gene (Gene ID: 20897) in murine species. STRA6 is a multi-pass membrane protein that acts as a high-affinity cell-surface receptor for the complex retinol-retinol binding protein (RBP/RBP4). It has one intramembrane and nine transmembrane helices in a dimeric assembly. It acts by removing retinol from RBP/RBP4 and transports it across the plasma membrane, where it can be metabolized. This mechanism does not depend on endocytosis. Binds to RBP/RBP4 with high affinity. Increases cellular retinol uptake from the retinol-RBP complex. STRA6 is widely expressed in the embryo and it in adults it is expressed choroid plexus and the brain microvascular cells that form the blood-organ barrier. It is also expressed in the retinal pigment epithelium, seminiferous epithelium in testis, the yolk sac, and in chorioallantoic placenta. Its expression in Sertoli cells depends on the stage of the spermatogenic cycle. STRA6 can be induced by retinoic acid and it is shown to be synergistically up-regulated by Wnt1 and retinoid in mammary epithelial cells. Mutations in Stra6 gene have been linked to the Matthew-Wood syndrome that is characterized by pulmonary agenesis/hypoplasia; microphthalmia/anophthalmia; and congenital cardiac, digestive, and urogenital malformations. (Ref.: Sadowski, S et al. (2017). Birth Defects Res. 109(4); 251-253; Chen, Y et al. (2016). Science 353 (6302), aad8266).

Specificity

This rabbit polyclonal antibody detects murine Stimulated by retinoic acid gene 6 protein (STRA6). it targets an epitope within 109 amino acids from the C-terminal region.

Immunogen

Epitope: C-terminus
His-tggged recombinant fragment corresponding to 109 amino acids from the C-terminal region of murine Stimulated by retinoic acid gene 6 (STRA6) protein.

Application

Anti-STRA6, Cat. No. ABN1662, is a highly specific rabbit polyclonal antibody that targets Stimulated by retinoic acid gene 6 protein (STRA6) and has been tested for use in Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.
Immunoprecipitation Analysis: A representative lot detected STRA6 in 293T cells transfected with an empty vector or pCMVTag2b-Stra6 (Courtesy of Dr. Noa Noy).

Western Blotting Analysis: A representative lot detected STRA6 in Western Blotting applications (Berry, D.C., et. al. (2012). Mol Cell Biol. 32(19):3851-9).

Western Blotting Analysis: A representative lot detected STRA6 in 293T cells transfected with an empty vector or pCMVTag2b-Stra6 (Courtesy of Dr. Noa Noy).

Flow Cytometry Analysis: A 1:100 dilution from a representative lot detected STRA6 in HT-29 cells.

Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected STRA6 in mouse testis and human liver cancer tissues.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in NIH3T3/L1Adipocytes.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected STRA6 in NIH3T3/L1 Adipocytes.

Target description

~90 kDa observed; 73.77 kda calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Unpurified
Rabbit polyclonal antiserum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bryan Marsh et al.
eLife, 9 (2020-11-04)
The placenta is the interface between mother and fetus in all eutherian species. However, our understanding of this essential organ remains incomplete. A substantial challenge has been the syncytial cells of the placenta, which have made dissociation and independent evaluation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service