ABN1379
Anti-TAG-1 Antibody
serum, from rabbit
Synonym(s):
Contactin-2, TAG-1, Axonal glycoprotein TAG-1, Axonin-1, Transient axonal glycoprotein 1, TAX-1
Select a Size
2.320,00 kr.
Select a Size
About This Item
2.320,00 kr.
Recommended Products
biological source
rabbit
Quality Level
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
rat, mouse
technique(s)
immunocytochemistry: suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
mouse ... Cntn2(21367)
rat ... Cntn2(25356)
General description
Immunogen
Application
Immunocytochemistry Analysis: A representative lot detected a loss of TAG-1 immunoreactivity at the juxtaparanodal (JXP) region of teased sciatic nerves isolated from 4.1B−/− mice lacking the cytoskeletal adapter protein 4.1B (Horresh, I., et al. (2010). J. Neurosci. 30(7):2480–2489).
Neuroscience
Developmental Neuroscience
Quality
Western Blotting Analysis: A 1:5,000 dilution of this antibody detected TAG-1 in 10 µg of mouse brain tissue lysate.
Target description
Physical form
Storage and Stability
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service