ABF1207
Anti-phospho-Syk (Tyr323)
from rabbit, purified by affinity chromatography
Synonym(s):
Tyrosine-protein kinase Syk, Tyr323 phosphorylated, p72-Syk, Tyr323 phosphorylated, p72Syk, Tyr323 phosphorylated, Spleen tyrosine kinase, Tyr323 phosphorylated
About This Item
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
phosphorylation (pTyr323)
Gene Information
human ... SYK(6850)
General description
Specificity
Immunogen
Application
Inflammation & Immunology
Quality
Western Blotting Analysis: A 1:5,000 dilution of this antibody detected Syk Tyr323 phosphorylation induction in 10 µg of lysate from hydrogen peroxide-treated Jurkat cells.
Target description
Linkage
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service