Skip to Content
Merck
All Photos(2)

Key Documents

ABE551

Sigma-Aldrich

Anti-E3 ubiquitin-protein ligase UHRF1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

E3 ubiquitin-protein ligase UHRF1, Inverted CCAAT box-binding protein of 90 kDa, Nuclear protein 95, Nuclear zinc finger protein Np95, HuNp95, hNp95, RING finger protein 106, Transcription factor ICBP90, Ubiquitin-like PHD and RING finger domain-containi

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
2.220,00 kr.

2.220,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
2.220,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

2.220,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

primate (based on 100% sequence homology), bovine (based on 100% sequence homology), porcine (based on 100% sequence homology), feline (based on 100% sequence homology), horse (based on 100% sequence homology), canine (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Ubiquitin-like with PHD and ring finger domains 1 (UHRF1), also known as ICBP90 or Np95, is a multifunctional nuclear protein that may be an important mediator of heterochromatin formation which is indicative of gene repression. UHRF1 contains a set and RING-finger-associated domain that detects hemi-methylated cytosine residues leading to the recruitment of DNMT1 to these residues during the S phase. UHRF1 thereby facilitates the transfer of methylation status to newly synthesized DNA molecules. UHRF1 may also interact with histones and HDAC1 via its PHD domain to promote modifications of histone residues which contribute to heterochromatin formation. UHRF1 may also directly modify histone H3 by ubiquitination. In addition, UHRF1 may contribute to the DNA-repair process though associations with PCNA. Several studies have suggested that overexpression of UHRF1 may contribute to the development of many cancers.

Immunogen

KLH-conjugated linear peptide corresponding to a region near the C-terminus of human E3 ubiquitin-protein ligase UHRF1.

Application

Detect UHRF1 using this rabbit polyclonal antibody, Anti-E3 ubiquitin-protein ligase UHRF1 Antibody validated for use in western blotting & IHC.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected E3 ubiquitin-protein ligase UHRF1 in human breast cancer tissue.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected E3 ubiquitin-protein ligase UHRF1 in 10 µg of HEK293 cell lysate.

Target description

~97 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lingyu Qiu et al.
Nucleic acids research, 51(17), 9166-9182 (2023-07-28)
Histone deacetylase 6 (HDAC6) mediates DNA damage signaling by regulating the mismatch repair and nucleotide excision repair pathways. Whether HDAC6 also mediates DNA double-strand break (DSB) repair is unclear. Here, we report that HDAC6 negatively regulates DSB repair in an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service