ABE328
Anti-dimethyl Histone H3 (Arg2) Antibody
from rabbit, purified by affinity chromatography
Synonym(s):
Histone H3.1, Histone H3/a, Histone H3/b, histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l
About This Item
Recommended Products
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
technique(s)
dot blot: suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
dimethylation (Arg2)
Gene Information
human ... HIST1H3F(8968)
General description
Specificity
Immunogen
Application
Dot Blot (Specificity) Analysis: A 1:620 dilution from a representative lot detected dimethyl Histone H3 (Arg2) peptides, but not unmethylated Histone H3 (Arg2) or other peptides corresponding to modified histones.
Absurance is a trademark of EMD Millipore, Inc.
Quality
Western Blotting Analysis: A 1:620 dilution from a representative lot detected dimethyl Histone H3 (Arg2) in 10 µg of HeLa acid extract.
Target description
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service