Skip to Content
Merck
All Photos(2)

Documents

ABE1456

Sigma-Aldrich

Anti-NR4A3

from rabbit, purified by affinity chromatography

Synonym(s):

Nuclear receptor subfamily 4 group A member 3, Mitogen-induced nuclear orphan receptor, Neuron-derived orphan receptor 1, Nuclear hormone receptor NOR-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

General description

Nuclear receptor subfamily 4 group A member 3 (UniProt: Q92570; also known as NR4A3, Mitogen-induced nuclear orphan receptor, Neuron-derived orphan receptor 1, Nuclear hormone receptor NOR-1) is encoded by the NR4A3 (also known as CHN, CSMF, MINOR, NOR1, TEC) gene (Gene ID: 8013) in human. NR4A3 acts as a transcriptional activator that binds to regulatory elements in promoter regions in a cell- and response element (target)-specific manner. It induces gene expression by binding as monomers to the NR4A1 response element (NBRE) 5′-AAAAGGTCA-3′ site and as homodimers to the Nur response element (NurRE) site in the promoter of their regulated target genes. Its DNA binding region is located within amino acids 289-364. Isoform alpha of NR4A3 is highly expressed in skeletal muscle and isoform beta is predominantly expressed in skeletal muscle. Lower expression levels have been reported in fetal brain and in placenta. NR4A3 plays a role in the regulation of proliferation, survival and differentiation of many different cell types and also in metabolism and inflammation. It is also shown to negatively regulate myeloid progenitor cell proliferation by repressing RUNX1 in a NBRE site-independent manner. NR4A3 activity is induced by inflammatory stimuli in endothelial cells through an NF-kappa-B-dependent transactivation of the NR4A3 promoter. NR4A3 is reported to modulate feeding behavior and is involved in energy balance by inhibiting the glucocorticoid-induced orexigenic neuropeptides AGRP expression. It also regulates glucose transport by regulating translocation of the SLC2A4 transporter to the cell surface. Mutations in NR4A3 gene have been linked to Ewing sarcoma, a highly malignant, metastatic, primitive small round cell tumor of bone and soft tissue.

Specificity

This rabbit polyclonal antibody detects Nuclear receptor subfamily 4 group A member 3 (NR4A3) in human and murine cells.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to 25 amino acids from the N-terminal region of human Nuclear receptor subfamily 4 group A member 3 (NR4A3).

Application

Anti-NR4A3, Cat. No. ABE1456, is a highly specific rabbit polyclonal antibody that detects Nuclear receptor subfamily 4 group A member 3 (NR4A3) and has been tested for use in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected NR4A3 in HeLa, A431, and NIH/3T3 cells.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected NR4A3 in 10 µg of HEK293 cell lysate.

Target description

~59 kDa observed; 68.23 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service