Skip to Content
Merck
All Photos(2)

Key Documents

ABE1337

Sigma-Aldrich

Anti-SETD6

from rabbit, purified by affinity chromatography

Synonym(s):

N-lysine methyltransferase SETD6, SET domain-containing protein 6

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
840,00 kr.
100 μG
2.090,00 kr.

840,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
840,00 kr.
100 μG
2.090,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

840,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... SETD6(79918)

General description

N-lysine methyltransferase SETD6 (UniProt: Q8TBK2; also known as SET domain-containing protein 6) is encoded by the SETD6 gene (Gene ID: 79918) in human. N-lysine methyltransferase SETD6 is a SET domain protein that specifically monomethylates lysine 310 of NF-kappa B-p65 that leads to down-regulation of NF-kappa B transcription factor activity. It can also monomethylate lysine 8 of H2AZ (H2AZK8me1). SETD6 is required for the maintenance of embryonic stem cell self-renewal. Primary urothelial cells and normal bladder tissue have almost undetectable SETD6 levels that are shown to increase in transformed urothelial cells and is further increased in bladder cancer cells and tissues. Overexpression of SETD6 in transformed urothelial cells increases their survival and colony formation. SETD6 is shown to interact with the oxidative stress sensor protein DJ1, which is required for Nrf2-dependent transcription of antioxidant target genes. Under basal conditions, SETD6 and DJ1 are associated at chromatin and SETD6 inhibits DJ1 activity, which leads to the repression of Nrf2-dependent transcription. However, under conditions of oxidative stress SETD6 mRNA and protein levels decline that leads to elevation in Nrf2 expression and diminish interaction between SETD6 and DJ1. Two isoforms of SETD6 have been described that are produced by alternative splicing. (Ref.: Mukherjee, N., et al. (2017). Oncotarget. 8(9); 15114-15125; Chen, A., et al. (2016). Biochim. Biophys. Acta. 1859(2):420-427).

Specificity

This rabbit polyclonal antibody detects N-lysine methyltransferase SETD6 in human cells. it targets an epitope within 18 amino acids from the N-terminal region.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to 18 amino acids from the N-terminal region of human N-lysine methyltransferase SETD6, isoform 1.

Application

Anti-SETD6, Cat. No. ABE1337, is a highly specific rabbit polyclonal antibody that targets N-lysine methyltransferase SETD6 and has been tested for use in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected SETD6 in HEK293 cells.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in human kidney tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected SETD6 in human kidney tissue lysate.

Target description

~53 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service