Skip to Content
Merck
All Photos(1)

Key Documents

AB19167

Sigma-Aldrich

Anti-MMP-2 Antibody, whole molecule

Chemicon®, from rabbit

Synonym(s):

72 kDa gelatinase, 72kD type IV collagenase, Gelatinase A, Matrix metalloproteinase-2, collagenase type IV-A, matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase), matrix metalloproteinase 2, matrix metalloproteinase 2 (g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

bovine, rat

species reactivity (predicted by homology)

human, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... MMP2(4313)

General description

MMPs have a common mode of activation, a conserved amino acid sequence in the putative metal-binding active site region and are inhibited by specific tissue inhibitors of metalloproteinases (TIMPs). These MMPs and TIMPs could be expressed by either the cancer of the stromal cells. There is a co-operation between tumor and stromal cells, in particular for the production of 72-kD type IV collagenase, involved in the disruption of basement membranes. A lack of TIMP-1 expression from invasive cancer cells could also contribute to matrix destruction.

Specificity

AB19167 recognizes protein at 72 kDa, identified as matrix metalloproteinase 2. MMP-2 is also known as 72 kDa collagenase IV or gelatinase A. It is synthesized as a 631 amino acid proenzyme that is activated by cleavage of the first 80 amino acids. AB19167 shows no cross-reaction with (pro) and active forms of other MMPs. Cytoplasmic localization.
Reactivity with other species has not been confirmed.

Immunogen

A synthetic peptide from the second half of human MMP-2.
Epitope: Whole molecule

Application

Detect MMP-2 using this Anti-MMP-2 Antibody, whole molecule validated for use in IH(P) & WB.
Immunohistochemistry (formalin/paraffin):
10-20 µg/mL of a previous lot was kept for 30 min at RT. No special pretreatment is required for staining formalin-fixed paraffin-embedded sections.

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
MMPs & TIMPs

Quality

Routinely evaluated by Western Blot on PC12 lysates.

Western Blot Analysis: 1:500 dilution of this lot detected MMP-2 on 10 µg of PC12 lysates.

Target description

72 kDa

Linkage

Replaces: 04-1048

Physical form

Format: Purified
Protein A chromatography
Purified rabbit polyclonal in buffer containing 10 mM PBS, pH 7.4, with 0.2% BSA and 0.09% sodium azide.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.

Analysis Note

Control
Positive control: Conditioned, serum-free medium from TPA-treated human fetal lung (HFL-1) cells. Placenta or bladder, breast, or ovarian carcinomas.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong-Yi Kuo et al.
Cell cycle (Georgetown, Tex.), 13(19), 3132-3142 (2014-12-09)
Promyelocytic leukemia protein (PML) is emerging as an important tumor suppressor. Its expression is lost during the progression of several types of cancer, including lung cancer. The EGF receptor (EGFR), a membrane-bound receptor tyrosine kinase, transduces intracellular signals responsible for
Qi Sun et al.
Frontiers in neuroscience, 10, 593-593 (2017-01-20)
This study was to explore the mechanisms underlying 1,2-dichloroethane (1,2-DCE) induced brain edema by focusing on alteration of matrix metalloproteinase-2 (MMP-2) in rat astrocytes induced by 2-chloroethanol (2-CE), an intermediate metabolite of 1,2-DCE in vivo. Protein and mRNA levels of
Silibinin inhibits established prostate tumor growth, progression, invasion, and metastasis and suppresses tumor angiogenesis and epithelial-mesenchymal transition in transgenic adenocarcinoma of the mouse prostate model mice.
Singh, RP; Raina, K; Sharma, G; Agarwal, R
Clinical cancer research : an official journal of the American Association for Cancer Research null
Marvin Ferrer et al.
Cell and tissue research, 349(3), 881-895 (2012-06-26)
Sperm-zona pellucida (ZP) penetration during fertilization is a process that most likely involves enzymatic digestion of this extracellular coat by spermatozoa. Since the inner acrosomal membrane (IAM) is the leading edge of spermatozoa during penetration and proteins required for secondary
Paul Stroemer et al.
Neurorehabilitation and neural repair, 23(9), 895-909 (2009-07-28)
This study investigated behavioral recovery in rats following implanting increasing doses of CTX0E03 cells into the putamen ipsilateral to the stroke damage. Postmortem histological analysis investigated possible mechanisms of behavioral recovery. . At 4 weeks after middle cerebral artery occlusion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service