Skip to Content
Merck
All Photos(1)

Key Documents

AB15624

Sigma-Aldrich

Anti-Tbx1 Antibody

Chemicon®, from rabbit

Synonym(s):

T-box 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
3.470,00 kr.

3.470,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
3.470,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

3.470,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

General description

The T-box 1 protein appears to be necessary for the normal development of large arteries that carry blood out of the heart, muscles and bones of the face and neck, and glands such as the thymus and parathyroid. Although the T-box 1 protein acts as a transcription factor, it is not yet known which genes are regulated by the protein.

Specificity

Other species have not been tested. The immunogen sequence has 76% homology with mouse.
Tbx1 (T-box 1).

Immunogen

Synthetic peptide from human Tbx1.

Application

Anti-Tbx1 Antibody is a Rabbit Polyclonal Antibody for detection of Tbx1 also known as T-box 1 & has been validated in WB.

Physical form

Format: Purified

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service