Skip to Content
Merck
All Photos(3)

Key Documents

AB10546

Sigma-Aldrich

Anti-Bcl-2-associated transcription factor 1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

BCL2-associated transcription factor 1, bcl-2-associated transcription factor 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
3.560,00 kr.

3.560,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
3.560,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

3.560,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), canine (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BCLAF1(9774)

General description

Bcl-2-associated transcription factor 1 (Btf) is a ubiquitously expressed death-promoting transcriptional repressor. It has been shown to interact with anti-apoptotic proteins related to Bcl-2, DNA, adenovirus E1B 19K, and EMD. Btf has also been shown to be involved in E1A transformation of p53 or E1B 19K, an interaction that implicates Btf as being involved in transcriptional regulation and in promoting apoptosis. Expression of Btf is absent in some cancers, which may suggest a potential role for the protein in tumor suppression.

Specificity

Other homologies: Rat (81% sequence homology).

Immunogen

KLH-conjugated linear peptide corresponding to human Bcl-2-associated transcription factor 1.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
BCL2 & Inhibition
This Anti-Bcl-2-associated transcription factor 1 Antibody is validated for use in WB, IP, IH(P) for the detection of Bcl-2-associated transcription factor 1.
Western Blot (Snap i.d.) Analysis: 0.5 µg/mL from a previous lot detected Bcl-2-associated transcription factor 1 in 10 µg of HeLa cell lysate.

Immunoprecipitation Analysis: 10 µg of this antibody immunoprecipitated Bcl-2-associated transcription factor 1 from HeLa cell lysate.

Immunohistochemistry Analysis: A 1:500 dilution from a previous lot detected Bcl-2-associated transcription factor 1 in neuronal and glial cells in normal human brain tissue.

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Bcl-2-associated transcription factor 1 in 10 µg of HeLa cell lysate.

Target description

~120 kDa observed.
This protein has 4 isoforms: Isoform 1 (106 kDa calculated), Isoform 2 (106 kda calculated), Isoform 3 (100 kDa calculated), Isoform 4 (86 kDa calculated).

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service