Skip to Content
Merck
All Photos(1)

Key Documents

8.45024

Sigma-Aldrich

Benzyl chloride

Synonym(s):

Benzyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7Cl
CAS Number:
Molecular Weight:
126.58
MDL number:
UNSPSC Code:
12352101

vapor pressure

1.2 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

autoignition temp.

585 °C

potency

440 mg/kg LD50, oral (Rat)

expl. lim.

1.1-14 % (v/v)

bp

179.3 °C/1013 hPa

mp

-39 °C

transition temp

flash point 67 °C

density

1.10 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C7H7Cl/c8-6-7-4-2-1-3-5-7/h1-5H,6H2

InChI key

KCXMKQUNVWSEMD-UHFFFAOYSA-N

Analysis Note

Colour (visual): colourless
Appearance of substance (visual): clear
Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.099 - 1.100
Identity (IR): passes test

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service