Skip to Content
Merck
All Photos(1)

Key Documents

8.43830

Sigma-Aldrich

1-Ethyl-1H-imidazole

for synthesis

Synonym(s):

1-Ethyl-1H-imidazole

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
578,00 kr.
100 ML
1.170,00 kr.

578,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
578,00 kr.
100 ML
1.170,00 kr.

About This Item

Empirical Formula (Hill Notation):
C5H8N2
CAS Number:
Molecular Weight:
96.13
MDL number:
UNSPSC Code:
12352005
EC Index Number:
230-403-9
NACRES:
NA.22

578,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

0.32 hPa ( 25 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

density

0.995 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[n]1(cncc1)CC

InChI

1S/C5H8N2/c1-2-7-4-3-6-5-7/h3-5H,2H2,1H3

InChI key

IWDFHWZHHOSSGR-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.993 - 0.997
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service