Skip to Content
Merck
All Photos(1)

Key Documents

8.41567

Sigma-Aldrich

2,5-Dimethylbenzothiazole

for synthesis

Synonym(s):

2,5-Dimethylbenzothiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NS
CAS Number:
Molecular Weight:
163.24
UNSPSC Code:
12352005
EC Index Number:
202-404-4

Quality Level

Assay

≥97.0% (GC)

form

crystals

potency

957 mg/kg LD50, oral (Rat)

mp

40-42 °C

storage temp.

2-30°C

InChI

1S/C9H9NS/c1-6-3-4-9-8(5-6)10-7(2)11-9/h3-5H,1-2H3

InChI key

XHANCLXYCNTZMM-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 39 °C
Melting range (upper value): ≤ 42 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service