Skip to Content
Merck
All Photos(1)

Key Documents

8.41331

Sigma-Aldrich

Pentafluorobenzoyl chloride

for synthesis

Synonym(s):

Pentafluorobenzoyl chloride, Perfluorobenzoyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
639,00 kr.
25 ML
2.460,00 kr.

639,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
639,00 kr.
25 ML
2.460,00 kr.

About This Item

Empirical Formula (Hill Notation):
C7ClF5O
CAS Number:
Molecular Weight:
230.52
MDL number:
UNSPSC Code:
12352106
EC Index Number:
218-842-4
NACRES:
NA.22

639,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

liquid

bp

158-159 °C/1013 hPa

density

1.664 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Fc1c(c(c(c(c1F)F)C(=O)Cl)F)F

InChI

1S/C7ClF5O/c8-7(14)1-2(9)4(11)6(13)5(12)3(1)10

InChI key

MYHOHFDYWMPGJY-UHFFFAOYSA-N

Application

  • GC-MS analysis of biological nitrate and nitrite using pentafluorobenzyl bromide in aqueous acetone: A dual role of carbonate/bicarbonate as an enhancer … A study on the non-catalytic role of carbonate/bicarbonate in derivatization reactions with pentafluorobenzoyl chloride (D Tsikas, 2021).
  • Photoconjugation of temperature-and pH-responsive polymer with silica nanoparticles for separation and enrichment of bacteria A method involving amidation reaction between amino-functionalized silica and pentafluorobenzoyl chloride (H Zheng, H Gong, L Cao, H Lin, L Ye, 2021).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.658 - 1.662
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service