8.24510
cis-Platinum
65% Pt, for synthesis
Synonym(s):
cis-Platinum (65% Pt), cis-Diaminedichloroplatinum(II), cis-Dichlorodiamineplatinum(II)
Select a Size
504,00 kr.
Select a Size
About This Item
504,00 kr.
Recommended Products
Product Name
cis-Platinum (65% Pt), for synthesis
Quality Level
potency
25.8 mg/kg LD50, oral (Rat)
mp
270 °C (decomposition)
solubility
2.5 g/L
density
3.7 g/cm3 at 20 °C
bulk density
850 kg/m3
storage temp.
2-30°C
InChI
1S/2Cl.2H3N.Pt/h;;2*1H3;
InChI key
JFUARQHXOQHNLK-UHFFFAOYSA-N
Analysis Note
Identity (IR): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Oral - Carc. 1B - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service