Skip to Content
Merck
All Photos(1)

Key Documents

8.14141

Sigma-Aldrich

Butyltriphenylphosphonium chloride

for synthesis

Synonym(s):

Butyltriphenylphosphonium chloride, Triphenylbutylphosphonium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
433,00 kr.

433,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
433,00 kr.

About This Item

Linear Formula:
(C6H5)3P+C4H9Cl-
CAS Number:
Molecular Weight:
354.85
MDL number:
UNSPSC Code:
12352107
EC Index Number:
236-443-3
NACRES:
NA.22

433,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

powder

potency

320 mg/kg LD50, oral (Rat)

mp

223-226 °C

transition temp

flash point >270 °C

storage temp.

2-30°C

SMILES string

[P+](CCCC)(c3ccccc3)(c2ccccc2)c1ccccc1.[Cl-]

InChI

1S/C22H24P.ClH/c1-2-3-19-23(20-13-7-4-8-14-20,21-15-9-5-10-16-21)22-17-11-6-12-18-22;/h4-18H,2-3,19H2,1H3;1H/q+1;/p-1

InChI key

MFIUDWFSVDFDDY-UHFFFAOYSA-M

Analysis Note

Assay (argentometric): ≥ 96.5 %
Melting range (lower value): ≥ 223 °C
Melting range (upper value): ≤ 226 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

518.0 °F

Flash Point(C)

> 270 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service