81177N
Probumin® Bovine Serum Albumin Polymer Enhanced Euro, 30% Solution
29-31 g/dL protein (Biuret), USA origin, Tested for IgG, Blood banking: suitable, Serology: suitable, enzyme immunoassay: suitable
Select a Size
Select a Size
About This Item
Recommended Products
Product Name
Probumin® Bovine Serum Albumin Polymer Enhanced Euro, 30% Solution, pack of 50 mL
description
0.1% sodium azide as preservtive
Contains sodium azide less than 0.1 g/dL
Quality Level
Assay
29-31 g/dL protein basis (biuret)
form
solution
shelf life
36 mo.
mol wt
~66 kDa
purified by
heat shock fractionation
packaging
pack of 50 mL
manufacturer/tradename
Probumin®
origin
USA origin
concentration
29-31 g/dL protein (Biuret)
technique(s)
blood typing: suitable ( )
blood typing: suitable
cell culture | mammalian: suitable
dilution
(Immunohematology)
(Serology)
impurities
Tested for IgG
pH
7.2-7.4
foreign activity
Bluetongue, none detected
Vesicular Stomatitis Virus, none detected
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service