Skip to Content
Merck
All Photos(1)

Key Documents

8.01546

Sigma-Aldrich

tert-Butylamine

for synthesis

Synonym(s):

tert-Butylamine, 2-Amino-2-methylpropane

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
265,00 kr.
500 ML
322,00 kr.
1 L
435,00 kr.
35 KG
9.520,00 kr.

265,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
265,00 kr.
500 ML
322,00 kr.
1 L
435,00 kr.
35 KG
9.520,00 kr.

About This Item

Linear Formula:
(CH3)3CNH2
CAS Number:
Molecular Weight:
73.14
MDL number:
UNSPSC Code:
12352116
EC Index Number:
200-888-1
NACRES:
NA.22

265,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

300 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

375 °C (DIN 51794)

potency

80 mg/kg LD50, oral (Rat)

expl. lim.

1.5-9.2 % (v/v)

pH

12 (20 °C, 100 g/L in H2O)

bp

44 °C

mp

-72 °C

transition temp

flash point -38 °C (DIN 51755 Part 1)

density

0.69 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

NC(C)(C)C

InChI

1S/C4H11N/c1-4(2,3)5/h5H2,1-3H3

InChI key

YBRBMKDOPFTVDT-UHFFFAOYSA-N

Application

  • Synthesis of gold nanoparticles: tert-Butylamine is employed in a one-step synthesis process to create alkanethiyl-stabilized gold nanoparticles, offering precise control over core size and monolayer functionality, enhancing the potential for nanotechnology applications in electronics and catalysis (Kay et al., 2023).
  • Catalytic activity of silver nanoparticles: The compound facilitates the synthesis of mesoionic N-heterocyclic carbene-tethered silver nanoparticles, which exhibit enhanced stability and significant catalytic activity, crucial for chemical transformations and pharmaceutical synthesis (Nazemi et al., 2023).
  • Improving colloidal stability: tert-Butylamine is used to improve the colloidal stability of temperature-sensitive poly(N-isopropylacrylamide) solutions, which are vital in biomedical applications such as drug delivery systems and tissue engineering scaffolds (Labuta et al., 2018).
  • Phosphate solubilisation: Research also includes its use in novel phosphate-solubilising bacteria isolated from sewage sludge, contributing to enhanced phosphate availability in environmental management and agriculture (Mo et al., 2019).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.692 - 0.694
Water (K. F.): ≤ 0.50 %
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-36.4 °F - closed cup

Flash Point(C)

-38 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service