Skip to Content
Merck
All Photos(1)

Key Documents

8.01068

Sigma-Aldrich

Aluminium bromide

anhydrous for synthesis

Synonym(s):

Aluminium bromide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
545,00 kr.
100 G
1.580,00 kr.
8010682500
29.820,00 kr.

545,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
545,00 kr.
100 G
1.580,00 kr.
8010682500
29.820,00 kr.

About This Item

Empirical Formula (Hill Notation):
AlBr3
CAS Number:
Molecular Weight:
266.69
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-779-7
NACRES:
NA.22
grade:
for synthesis
form:
crystals

545,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

grade

for synthesis

Quality Level

vapor pressure

1.3 hPa ( 81 °C)

form

crystals

potency

1598 mg/kg LD50, oral (Rat)

bp

250-270 °C/1013 hPa

mp

97.5 °C

density

3.2 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Al+3].[Br-].[Br-].[Br-]

InChI

1S/Al.3BrH/h;3*1H/q+3;;;/p-3

InChI key

PQLAYKMGZDUDLQ-UHFFFAOYSA-K

Analysis Note

Assay (complexometric): ≥ 98.0 %
Identity (ICP-OES): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service