Skip to Content
Merck
All Photos(1)

Key Documents

8.00668

Sigma-Aldrich

Pimelic acid

for synthesis

Synonym(s):

Pimelic acid, Heptanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOC(CH2)5COOH
CAS Number:
Molecular Weight:
160.17
MDL number:
UNSPSC Code:
12352106
EC Index Number:
203-840-8

Assay

≥98.0% (GC)

form

crystals

potency

7000 mg/kg LD50, oral (Rat)

bp

404 °C/1013 hPa

mp

102-105 °C

solubility

50 g/L

density

1.329 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C7H12O4/c8-6(9)4-2-1-3-5-7(10)11/h1-5H2,(H,8,9)(H,10,11)

InChI key

WLJVNTCWHIRURA-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 103 °C
Melting range (upper value): ≤ 107 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service