Skip to Content
Merck
All Photos(1)

Key Documents

78900BC

Millipore

Gemini® 2-Lane Sample Rack

BioControl, For use with Gemini®

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
11.260,00 kr.

11.260,00 kr.


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 UNIT
11.260,00 kr.

About This Item

UNSPSC Code:
41122409

11.260,00 kr.


Please contact Customer Service for Availability

Request more information

packaging

pkg of 1 unit

manufacturer/tradename

BioControl Systems 78900
Gemini®

storage condition

room temperature

technique(s)

ELISA: suitable
enzyme immunoassay: suitable

application(s)

food and beverages
pathogen testing

compatibility

BioControl Gemini

storage temp.

room temp

Components

One sample rack

Testing Method

ELISA

Legal Information

Gemini is a registered trademark of Stratec

related product


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service