Skip to Content
Merck
All Photos(1)

Key Documents

740

Sigma-Aldrich

Wright Stain

Harleco®, suitable for hematology

Synonym(s):

Hematology Stain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121
EC Index Number:
200-659-6

form

liquid

Quality Level

manufacturer/tradename

Harleco®

mp

-97.77 °C

density

0.792 g/cm3

λmax

520-530 nm (Eosin)
645-655 nm (Azure)

application(s)

diagnostic assay manufacturing
hematology ((contains Methanol))

shipped in

ambient

storage temp.

15-25°C

General description

As a gold standard in hematology, the Harleco® Wright stain is a Romanowsky neutral dye stain based on the combination of methylene blue azure as the basic dye component and eosin as the acidic dye component. The typical purple color of cell nuclei is due to a molecular interaction between eosin Y and an azure B-DNA complex.

This polychromic stain is widely used to identify acidophilic, basophilic, and neutrophilic granules of leukocytes in peripheral blood and bone marrow smears. For decades, the Harleco Wright stain has been used in clinical diagnostics to characterize red blood cell morphology and to differentiate white blood cell types. The staining result can be influenced by several factors such as pH, buffer composition, fixation, and staining time.

The Harleco Wright stain can be used to stain the following sample materials: fresh, native whole blood or bone-marrow smears and clinical cytology specimens (urine sediment, sputum, smears from fine needle aspiration biopsies (FNAB), rinses, and imprints). It may also be used for G-banding to aid in the analysis of poorly spread metaphases.
HARLECO® Polychromic stain for peripheral blood and bone marrow smears. Used for red blood cell morphology and white blood cell differentiation.

Application

HARLECO® Polychromic stain for peripheral blood and bone marrow smears. Used for red blood cell morphology and white blood cell differentiation.
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Polychromic stain for peripheral blood and bone marrow smears. Used for red blood cell morphology and white blood cell differentiation.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service