Skip to Content
Merck
All Photos(1)

Documents

70045-M

Sigma-Aldrich

pBAC4x-1 DNA - Novagen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106612
NACRES:
NA.51

form

liquid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

storage temp.

−20°C

General description

pBAC4x-1 is a baculovirus transfer plasmid designed for cloning and co-expression of up to four target genes or multiple copies of the same gene in insect cells. The plasmid contains two polh promoters and two p10 promoters, each of which is upstream of unique cloning sites for sequential insertion of target genes. The homologous promoters are in opposite orientations to minimize recombination. The plasmid is compatible with BacVector-1000, -2000 or -3000 Triple Cut Virus DNA for low background transfection and efficient utilization of the promoters. The vector provides optional expression of a C-terminal His•Tag fusion sequence from the polh right-hand promoter by allowing read-through of inserts in the proper reading frame. Unique restriction sites are indicated on the circle map. The cloning/expression region of the coding strand transcribed from the clockwise promoters is shown below. The f1 origin is oriented so that infection with helper phage will produce virions containing single stranded DNA that corresponds to this strand. Single stranded sequencing of phage-derived DNA can be performed using the 1629DWN primer.

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service