Skip to Content
Merck
All Photos(1)

Key Documents

6701

Millipore

Uridine-5′-triphosphate, Trisodium Salt

Synonym(s):

Uridine-5′-triphosphate, Trisodium Salt, UTP, 3Na

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
662,00 kr.

662,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
662,00 kr.

About This Item

Empirical Formula (Hill Notation):
C9H12N2O15P3 · 3Na
CAS Number:
Molecular Weight:
550.09
MDL number:
UNSPSC Code:
12352208
NACRES:
NA.51

662,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥97% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[P](=O)([O-])([O-])O[P](=O)([O-])O[P](=O)([O-])OC[C@H]1O[C@H]([C@@H]([C@@H]1O)O)N2C=CC(=O)NC2=O

InChI

1S/C9H15N2O15P3/c12-5-1-2-11(9(15)10-5)8-7(14)6(13)4(24-8)3-23-28(19,20)26-29(21,22)25-27(16,17)18/h1-2,4,6-8,13-14H,3H2,(H,19,20)(H,21,22)(H,10,12,15)(H2,16,17,18)/p-4/t4-,6-,7-,8-/m1/s1

InChI key

PGAVKCOVUIYSFO-XVFCMESISA-J

General description

Energy-rich precursor in the enzymatic biosynthesis of RNA. A potent vasodilator. Induces contractile responses in some tissues.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service