Skip to Content
Merck
All Photos(1)

Key Documents

6.18598

Sigma-Aldrich

Diethylamine

for chromatography

Synonym(s):

Diethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NH
CAS Number:
Molecular Weight:
73.14
MDL number:
UNSPSC Code:
12352116

grade

for chromatography

Quality Level

SMILES string

N(CC)CC

InChI

1S/C4H11N/c1-3-5-4-2/h5H,3-4H2,1-2H3

InChI key

HPNMFZURTQLUMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethylamine (DEA) is commonly used as an ​eluent additive in various chromatographic techniques, including reversed-phase high-performance liquid chromatography (HPLC), high pH reversed-phase preparative chromatography, and hydrophilic liquid chromatography (HILIC). It plays a crucial role in system efficiency and enhancing peak shape when used in column chromatography.

Application

Diethylamine can be used as a mobile phase modifierin the HILIC method for the simultaneous determination of paracetamol andmethionine in a soft capsule formulation.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separation of nadolol racemates by high pH reversed-phase preparative chromatography
Arafah RS, et al.
Separation and Purification Technology, 233, 116018-116018 (2020)
RP-HPLC ANALYSIS OF ACIDIC AND BASIC DRUGS IN SYSTEMS WITH DIETHYLAMINE AS ELUENTS ADDITIVE.
Petruczynik A, et al.
Acta Poloniae Pharmaceutica, 73(6), 1475-1485 (2016)
Peak shape enhancement using diethylamine in hydrophilic liquid interaction chromatography: Application in simultaneous determination of methionine and paracetamol
BinhVN, et al.
Journal of Pharmaceutical and Biomedical Analysis, 203, 114214-114214 (2021)
A new analytical methodology for a fast evaluation of semi-volatile polycyclic aromatic hydrocarbons in the vapor phase downstream of a diesel engine particulate filter
Journal of Chromatography A, 1218(7), 981-989 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service