6.18583
Tetrabutylammonium hydroxide solution
0.1 N in toluene/methanol
Synonym(s):
Tetrabutylammonium hydroxide solution
About This Item
Recommended Products
form
solution
concentration
0.1 N in toluene/methanol
SMILES string
[OH-].CCCC[N+](CCCC)(CCCC)CCCC
InChI
1S/C16H36N.H2O/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H2/q+1;/p-1
InChI key
VDZOOKBUILJEDG-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Application
- Supramolecular polymer networks: Tetrabutylammonium hydroxide serves as a crucial component in the synthesis of highly robust supramolecular polymer networks, crosslinked by metallacycles, offering potential advancements in materials science and engineering (Li et al., 2024).
- Biomedical hydrogel applications: This compound facilitates the development of pH-responsive cellulose/silk/Fe(3)O(4) hydrogel microbeads, designed for targeted drug delivery and other biomedical applications, demonstrating its versatility in healthcare innovations (Lee et al., 2024).
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 2
Target Organs
Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service