Skip to Content
Merck
All Photos(1)

Key Documents

6.17691

Sigma-Aldrich

Hexane

for chromatography

Synonym(s):

Hexane, n-Hexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH3
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
MDL number:
UNSPSC Code:
12352001

product name

n-Hexane, for chromatography

grade

for chromatography

Quality Level

vapor density

~3 (vs air)

vapor pressure

256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

autoignition temp.

453 °F

expl. lim.

7.7 %

refractive index

n20/D 1.375 (lit.)

bp

69 °C (lit.)

mp

−95 °C (lit.)

density

0.659 g/mL at 25 °C (lit.)

SMILES string

CCCCCC

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

n-Hexaneis typically used as a mobile phase solvent in normal-phase chromatography oras a diluent in reversed-phase chromatography. It can effectively separatecompounds that have similar chemical properties, such as hydrocarbons, fattyacids, and lipid samples.

Application

n-Hexanecan be used as a solvent for extraction of polycyclic aromatic hydrocarbons(PAHs) from smoked, grilled, and boiled meats using gas chromatography - flameionization detector (GC-FID).

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atmospheric pressure photoionization mass spectrometry for analysis of fatty acid and acylglycerol lipids
Cai SS, et al.
Journal of Chromatography A, 110(1-2), 15-26 (2006)
Determination of polycyclic aromatic hydrocarbons [PAHs] in processed meat products using gas chromatography--Flame ionization detector
Olatunji OS, et al.
Food Chemistry, 156, 296-300 (2014)
Alternative solvents for improving the greenness of normal phase liquid chromatography of lipid classes
Prache N, et al.
Journal of Chromatography A, 1464, 55-63 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service