Skip to Content
Merck
All Photos(1)

Key Documents

581810

Sigma-Aldrich

(-)-Terreic Acid, Synthetic

a cell-permeable quinone epoxide antibiotic

Synonym(s):

(-)-Terreic Acid, Synthetic, (1R,6S)-3-Hydroxy-4-methyl-7-oxabicyclo[4.1.0]hept-3-ene-2,5-dione, BTK Inhibitor II, Bruton′s Tyrosine Kinase Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6O4
CAS Number:
Molecular Weight:
154.12
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

solubility

water: 5 mg/mL

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C7H6O4/c1-2-3(8)5(10)7-6(11-7)4(2)9/h6-8H,1H3/t6-,7+/m1/s1

InChI key

ATFNSNUJZOYXFC-RQJHMYQMSA-N

General description

A cell-permeable quinone epoxide antibiotic that acts as a reversible, substrate competitive, and selective inhibitor of Bruton′s tyrosine kinase (BTK; IC50 = 10 µM and 3 µM for the basal and activation levels), both in vitro and in vivo. Shown to bind to the BTK pleckstrin homology domain (BTK-PH) and block the interaction between BTK-PH and protein kinase C (PKC) (IC50 ~100 µM in human mast cell lysates), thus affecting the catalytic activity of BTK, but not the activity of PKC. Selectively inhibits the autophosphorylation of PKCβI (IC50 ~8 µM) but not that of PKCβII. Reported to effectively inhibit the production of TNF-α (IC50 ~3 µM) and block the activation of JNK1 (IC50 ~10 µM) in FcεRI-stimulated mast cells. Also reported to inhibit DNA synthesis in activated spleen cells (IC50 ~1.5 µM). Does not significantly affect the activities of Lyn, Syk, PKA, CK-1, ERK1, ERK2 and p38 kinases.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Bruton′s tyrosine kinase catalytic activity (BTK)
Product competes with ATP.
Reversible: yes
Target IC50: 10 µM against BTK

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kawakami, Y., et al. 2000. Proc. Natl. Acad. Sci. USA97, 7423.
Kawakami, Y., et al. 1999. Proc. Natl. Acad. Sci. USA96, 2227.
Yamamoto, H., et al. 1980. Jpn. J. Antibiot.33, 320.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service