Skip to Content
Merck
All Photos(1)

Key Documents

5.33574

Sigma-Aldrich

TRPML3 Activator, ML123

Synonym(s):

TRPML3 Activator, ML123, 4-Chloro-N-(2-morpholinocyclohexyl)benzenesulfonamide, SF-21, CID2911646, ML123, ML 123, Transient Receptor Potential Channel 3 Activator, SID24801657, Racemate, SR-1984

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H23ClN2O3S
CAS Number:
Molecular Weight:
358.88
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Product 5.33574 is not currently sold in your country. Contact Technical Service

Assay

≥92% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

C1CCC(C(C1)NS(=O)(=O)C2=CC=C(C=C2)Cl)N3CCOCC3

InChI key

AYQCZQKMGKDZPS-UHFFFAOYSA-N

General description

A non-cytotoxic morpholinyl-cyclohexylbenzenesulfonamide compound that acts as a selective agonist of TRPML3 channel (EC50 = 873 nM in hTRPML3-YPF transfected HEK293 cells). Displays significantly weaker agonistic activity for TRPML2 channel (54.5% at 10 µM) and is shown to be inactive against hTRPML1, hTRPM2, mTRPV2, hTRPC3, drTRPN1, and hTRPA1 (~10 µM). Also binds to sigma-1 receptor at much higher concentrations (IC50 = 31 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
TRPML3
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Antolin, A.A., and Mestres, J. 2015. ACS Chem. Biol.10, 395.

Grimm, C., et al. 2012. J. Biol. Chem.287, 22701.
Saldanha, S., et al. 2010. Probe Reports from the NIH Molecular Libraries Program.
Grimm, C., et al. 2010. Chem. Biol.17, 135.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service