Skip to Content
Merck
All Photos(1)

Key Documents

5.30656

Sigma-Aldrich

LTA4 Epoxide Hydrolase Inhibitor, ARM1

Synonym(s):

LTA4 Epoxide Hydrolase Inhibitor, ARM1, 4-(4-Benzylphenyl)-thiazol-2-amine, Leukotriene A4 Epoxide Hydrolase Inhibitor, LTA₄ Hydrolase Inhibitorq, LTA4H Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14N2S
CAS Number:
Molecular Weight:
266.36
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

C1=CC=C(C=C1)CC2=CC=C(C=C2)C3=CSC(=N3)N

General description

A cell-permeable benzylphenyl-thiazolamine compound that selectively inhibits against the leukotriene (LT) A4 epoxide hydrolase activity of LTA4H by occupying the same hydrophobic tunnel (Ki = 2.3 µM) targeted by LTA4 ω-end, displaying much reduced potency against LTA4H aminopeptidase activity (% inhibition/[ARM1] = 0/100 µM & 35/1 mM; Substrate = 800 µM Pro-Gly-Pro) coordinated by E296 and the catalytic site Zn2+ at the adjacent hydrophilic peptide substrate-binding cavity. Shown to inhibit A23187- (Cat. Nos. 100105 & 100106) induced LTB4 production in primary human polymorphonuclear neutrophil (PMN) cultures (IC50 = 0.5 µM; ARM1 added 10 min prior to 5 min stimulation by 2.5 µM A23187). Another LTA4H inhibitor SC 57461A (CAS 423169-68-0), in comparison, inhibits both the LTA4-to-LTB4 conversion as well as the chemotactic tripeptide Pro-Gly-Pro degradation activity of LTA4H.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
LTA4
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Use only fresh DMSO for reconstitution.

Other Notes

Stsiapanava, A., et al. 2014. Proc. Natl. Acad. Sci. USA111, 4227.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service