Skip to Content
Merck
All Photos(1)

Key Documents

5.30537

Sigma-Aldrich

JmjC Histone Demethylase Inhibitor, n-Octyl-IOX1

Synonym(s):

JmjC Histone Demethylase Inhibitor, n-Octyl-IOX1, KDM4 Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
1.650,00 kr.

1.650,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
10 MG
1.650,00 kr.

About This Item

Empirical Formula (Hill Notation):
C18H23NO3
CAS Number:
Molecular Weight:
301.38
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

1.650,00 kr.


Please contact Customer Service for Availability

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

gray-green

solubility

DMSO: 5 mg/mL

storage temp.

−20°C

General description

A cell-permeable, non-toxic, n-octyl ester form of 5-carboxy-8-hydroxyquinoline (IOX1, Cat. No. 420201) that acts as a superior inhibitor of Jumonji C domain histone lysine demethylase. Shown to be about 30 fold more potent than IOX1 (EC50 = 3.8 µM for KDM4A in HeLa cells vs. 100 µM for IOX1). Inhibits the H3K9 me3 demethylation activity of KDM4C (IC50 = 3.9 µM in ALPHA Screen assay). Exhibits greater stability to hydrolysis in the ALPHA Screen buffer.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
KDM4
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Schiller, R., et al. 2014. ChemMedChem.9,566.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service