529584
PTK/PI 3-K/mTOR Inhibitor, PP121
The PTK/PI 3-K/mTOR Inhibitor, PP121, also referenced under CAS 1092788-83-4, controls the biological activity of PTK/PI 3-K/mTOR. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Synonym(s):
PTK/PI 3-K/mTOR Inhibitor, PP121, 1-Cyclopentyl-3-(1H-pyrrolo[2,3-b]pyridin-5-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine, PI 3-K/mTOR Inhibitor II, PDGFR Tyrosine Kinase Inhibitor XVIII, Abl Inhibitor IV, mTOR Inhibitor VII, VEGFR Tyrosine Kinase Inhibitor XXXII, VEGFR2 Kinase Inhibitor XXX, PI 3-K Inhibitor XIII
Select a Size
1.150,00 kr.
Select a Size
About This Item
1.150,00 kr.
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white
solubility
DMSO: 100 mg/mL
shipped in
ambient
storage temp.
−20°C
SMILES string
[n]2(nc(c5c2ncnc5N)c3cnc4[nH]ccc4c3)C1CCCC1
InChI
1S/C17H17N7/c18-15-13-14(11-7-10-5-6-19-16(10)20-8-11)23-24(12-3-1-2-4-12)17(13)22-9-21-15/h5-9,12H,1-4H2,(H,19,20)(H2,18,21,22)
InChI key
NVRXTLZYXZNATH-UHFFFAOYSA-N
General description
Packaging
Warning
Reconstitution
Other Notes
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service