Skip to Content
Merck
All Photos(1)

Key Documents

5.09522

Sigma-Aldrich

ERW1041E

≥98% (HPLC), solid, TG2 inhibitor, Calbiochem®

Synonym(s):

Transglutaminase 2 Inhibitor II, ERW1041E, TG2 Inhibitor, TGM2 Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
2.000,00 kr.

2.000,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
2.000,00 kr.

About This Item

Empirical Formula (Hill Notation):
C20H21BrN4O4
CAS Number:
Molecular Weight:
461.31
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

2.000,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Transglutaminase 2 Inhibitor II, ERW1041E,

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

11 μM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

−20°C

SMILES string

C1CC(N(C1)C(=O)OCC2=CC3=CC=CC=C3N=C2)C(=O)NCC4CC(=NO4)Br

General description

A cell-permeable, dihydroisoxazole compound that acts as an active-site directed, acyl-donor substrate competitive inhibitor of transglutaminase 2 (TG2, Ki = 11 µM). Shown to bind to the active site cysteine. Blocks 5-biotinylamido pentylamine (5BP)-induced TG2 activation in C57BL/6 mice lung in a dose-dependent manner and reduces hypoxia-induced pulmonary hypertension. Blocks poly(I:C)-induced TG2 activity in small intestine without affecting villous atrophy. Exhibits good selectivity over TG3 and Factor XIIIa, but has equipotent effect on TG1. It is well-tolerated by mice (~50 mg/kg) and displays moderate pharmacokinetic properties with t1/2= 12 h.
A cell-permeable, dihydroisoxazole compound that acts as an active-site directed, acyl-donor substrate competitive inhibitor of transglutaminase 2 (TG2, Ki = 11 µM). Shown to bind to the active site cysteine. Blocks 5-biotinylamido pentylamine (5BP)-induced TG2 activation in C57BL/6 mice lung in a dose-dependent manner and reduces hypoxia-induced pulmonary hypertension. Blocks poly(I:C)-induced TG2 activity in small intestine without affecting villous atrophy. Exhibits good selectivity over TG3 and Factor XIIIa, but has equipotent effect on TG1. It is well-tolerated by mice (~50 mg/kg) and displays moderate pharmacokinetic properties with t1/2= 12 h.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Primary Target
TGM2

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

DiRaimond, T.R., et al. 2014. ACS Chem. Biol.9, 266.
Dafik, L., et al. 2012. PLoS One.7, e30642.
Watts, R.W., et al. 2006. J. Med. Chem.49, 7493.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service