Skip to Content
Merck
All Photos(1)

Documents

5.08340

Sigma-Aldrich

PERK Inhibitor I, GSK2606414

GSK2606414

Synonym(s):

InSolution PERK Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H20F3N5O
CAS Number:
Molecular Weight:
451.44
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

SMILES string

O=C(CC1=CC=CC(C(F)(F)F)=C1)N2CCC3=C2C=CC(C4=CN(C)C5=C4C(N)=NC=N5)=C3

InChI

1S/C24H19F4N5O/c1-32-11-18(21-22(29)30-12-31-23(21)32)14-2-3-19-15(9-14)4-5-33(19)20(34)8-13-6-16(24(26,27)28)10-17(25)7-13/h2-3,6-7,9-12H,4-5,8H2,1H3,(H2,29,30,31)

InChI key

PXVQGBJMIQCDEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Glass bottle

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service