Skip to Content
Merck
All Photos(1)

Documents

481414

Sigma-Aldrich

NF-κB Activation Inhibitor VI, BOT-64 - CAS 113760-29-5 - Calbiochem

The NF-κB Activation Inhibitor VI, BOT-64, also referenced under CAS 113760-29-5, controls the biological activity of NF-κB. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

Synonym(s):

NF-κB Activation Inhibitor VI, BOT-64 - CAS 113760-29-5 - Calbiochem, 6,6-Dimethyl-2-(phenylimino)-6,7-dihydro-5H-benzo[1,3]oxathiol-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15NO2S
CAS Number:
Molecular Weight:
273.35
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

ethanol: 10 mg/mL
DMSO: 50 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable benzoxathiole compound that acts as an IKK-2 inhibitor, presumably by targeting the Ser177 and/or Ser181 residues in the kinase′s activation loop domain. Shown to inhibit LPS-induced NF-κB activation and nitrite production (IC50 = 1.0 & 0.7 µM, respectively) in RAW 264.7 cells in vitro and effectively prevent LPS-induced septic death in mice (70% survival rate with 30 mg/kg BOT-64, i.p.) in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Kim, B.H., et al. 2008. Mol. Pharmacol.73, 1309.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Byung Hak Kim et al.
Molecular pharmacology, 73(4), 1309-1318 (2008-01-19)
Benzoxathiole derivatives have been used in the treatment of acne and have shown cytostatic, antipsoriatic, and antibacterial properties. However, little is known about the molecular basis for these pharmacological properties, although nuclear factor (NF)-kappaB activation is closely linked to inflammation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service