Skip to Content
Merck
All Photos(1)

Key Documents

420138

Sigma-Aldrich

InhibitorSelect JAK/STAT Signaling Pathway Inhibitor Panel

The InhibitorSelect JAK/STAT Signaling Pathway Inhibitor Panel controls the biological activity of InhibitorSelect.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

shipped in

wet ice

storage temp.

−20°C

General description

A convenient panel containing 13 potent and selective inhibitors useful for the study of JAK/STAT signaling pathway. This panel contains the following inhibitors as well as 15 ml anhydrous DMSO for reconstitution:
A convenient panel containing 13 potent and selective inhibitors useful for the study of JAK/STAT signaling pathway. This panel contains the following inhibitors: 1 mg of Indirubin Derivative E804 (Cat. No. 402081); 500 µg of JAK Inhibitor I (Cat. No. 420099); 25 mg of JAK2 Inhibitor II (Cat. No. 420132); 5 mg of LY 294002 (Cat. No. 440202); 1 mg of PD 153035 (Cat. No. 234490); 1 mg of PP2 (Cat. No. 529573); 50 mg of SHP1/2 PTPase Inhibitor, NSC-87877 (Cat. No. 565851); 1 mg of STAT3 Inhibitor Peptide, Cell-Permeable (Cat. No. 573096); 10 mg of STAT3 Inhibitor III, WP1066 (Cat. No. 573097); 25 mg of STAT3 Inhibitor V, Stattic (Cat. No. 573099); 10 mg of STAT5 Inhibitior (Cat. No. 573108); 5 mg of AG490 (Cat. No. 658401) and 1 mg of U0126 (Cat. No. 662005). Also provided with 15 ml of anhydrous DMSO.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) each inhibitor. Each inhibitor solution is stable for up to 3 months at -20°C.
Please refer to information provided on the CD for reconstitution information of individual inhibitors.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service