Skip to Content
Merck
All Photos(1)

Key Documents

3810-OP

Millipore

DTE

OmniPur® Grade, >= 99.0%

Synonym(s):

OmniPur® DTE, Dithioerythitol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

Quality Level

form

crystalline powder

reaction suitability

reagent type: reductant

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

pH

4.5-6.5 (25 °C, 15.4 g/L)

foreign activity

DNAse, none detected
Protease, none detected
RNAse, none detected

shipped in

ambient

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4+

InChI key

VHJLVAABSRFDPM-ZXZARUISSA-N

General description

Molecular biology grade.

Specifications:
A (285 nm, 0.02 M, water): ≤0.15AU
DNase: None detected
Melting point: 82-85°C
Protease: None detected
Purity: ≥99.0%
RNase: None detected
Solubility (5%, water): To pass test





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service