Skip to Content
Merck
All Photos(1)

Key Documents

373402

Sigma-Aldrich

Hedgehog Antagonist VIII

The Hedgehog Antagonist VIII, also referenced under CAS 330796-24-2, controls the biological activity of Hedgehog. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

Hedgehog Antagonist VIII, 1-(4-Chloro-3-(trifluoromethyl)phenyl)-3-((3-(4-fluorophenyl)-3,4-dihydro-4-oxo-2-quinazolinyl)methyl)-urea, Hh Signaling Antagonist VIII, Hh Signaling Antagonist VIII, 1-(4-Chloro-3-(trifluoromethyl)phenyl)-3-((3-(4-fluorophenyl)-3,4-dihydro-4-oxo-2-quinazolinyl)methyl)-urea

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
1.310,00 kr.

1.310,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
5 MG
1.310,00 kr.

About This Item

Empirical Formula (Hill Notation):
C23H15ClF4N4O2
CAS Number:
Molecular Weight:
490.84
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

1.310,00 kr.


Please contact Customer Service for Availability

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

clear slightly yellow

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

FC(F)(F)c1c(ccc(c1)NC(=O)NCc2[n]([c](c4c(n2)cccc4)=O)c3ccc(cc3)F)Cl

InChI

1S/C23H15ClF4N4O2/c24-18-10-7-14(11-17(18)23(26,27)28)30-22(34)29-12-20-31-19-4-2-1-3-16(19)21(33)32(20)15-8-5-13(25)6-9-15/h1-11H,12H2,(H2,29,30,34)

InChI key

NYOXUHUPXGFHIB-UHFFFAOYSA-N

General description

A cell-permeable quinazolinyl-urea compound that is shown to potently inhibit OCT-Shh-stimulated Gli transcription activity in a 10t1/2(s12) cell-based Luciferase reporter assay (IC50 = 70 nM).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C0. Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Brunton, A.A., et al. 2008. J. Med. Chem.51, 1108.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service