Skip to Content
Merck
All Photos(1)

Key Documents

346125

Millipore

D-Gluconic Acid, Potassium Salt

D-Gluconic acid is an aldonic acid formed by the oxidation of the carbonyl carbon of glucose in microorganisms, plants, and animals. In solution, it exists as an equilibrium mixture of free acid and lactone.

Synonym(s):

D-Gluconic Acid, Potassium Salt, Potassium D-Gluconate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11O7 · K
CAS Number:
Molecular Weight:
234.25
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.79

Quality Level

Assay

≥99% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

15-25°C

InChI

1S/C6H12O7.K/c7-1-2(8)3(9)4(10)5(11)6(12)13;/h2-5,7-11H,1H2,(H,12,13);/q;+1/p-1/t2-,3-,4+,5-;/m1./s1

InChI key

HLCFGWHYROZGBI-JJKGCWMISA-M

General description

D-Gluconic acid is an aldonic acid formed by the oxidation of the carbonyl carbon of glucose in microorganisms, plants, and animals. In solution, it exists as an equilibrium mixture of free acid and lactone.
In solution, exists as an equilibrium mixture of free acid and lactone. D-Gluconic acid is an aldonic acid, a metabolite intermediate in microorganisms, plants, and animals.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Bausch, C., et al. 1998. J. Bacteriol. 180, 3704.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

C Bausch et al.
Journal of bacteriology, 180(14), 3704-3710 (1998-07-11)
The presence of two systems in Escherichia coli for gluconate transport and phosphorylation is puzzling. The main system, GntI, is well characterized, while the subsidiary system, GntII, is poorly understood. Genomic sequence analysis of the region known to contain genes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service