Skip to Content
Merck
All Photos(1)

Key Documents

344301

Sigma-Aldrich

sFRP Inhibitor II

The sFRP Inhibitor II, also referenced under CAS 302804-57-5, controls the biological activity of sFRP. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

Synonym(s):

sFRP Inhibitor II, Wnt Pathway Activator V, 2-(2-(2-Imino-4-oxo-thiazolidin-5-yl)-acetylamino)-4,5-dimethyl-thiophene-3-carboxylic acid ethyl ester, Secreted Frizzled Related Protein-1 Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17N3O4S2
CAS Number:
Molecular Weight:
355.43
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable iminooxothiazolidine compound that is shown to counteract the antagonizing effect of SARP-2/sFRP-1 (secreted frizzled related protein-1) against Wnt-3/frizzled interaction and restore cellular response to Wnt-3 stimulation (by a 2.3-fold enhancement at 15 µM) in U2OS-based reporter assays. Also reported to increase total bone area and the number of osteoblasts (by ~20% and ~45%, respectively) in a murine ex vivo calvarial bone formation assay.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Shi, M., et al. 2009. Bioorg. Med. Chem. Lett.19, 6337.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service